Product Details

SNP ID
rs151147398
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:153907841 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTTCCTGCCCAGGCGGCTGCTGGGC[A/G]GGGCCTTTGGGCTTCGGGGCCCAGG
Phenotype
MIM: 300023 MIM: 300538
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ARHGAP4 PubMed Links

Gene Details

Gene
ARHGAP4
Gene Name
Rho GTPase activating protein 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001164741.1 2907 Missense Mutation CCG,CTG P950L NP_001158213.1
NM_001666.4 2907 Missense Mutation CCG,CTG P910L NP_001657.3
Gene
AVPR2
Gene Name
arginine vasopressin receptor 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000054.4 2907 Intron NP_000045.1
NM_001146151.1 2907 Intron NP_001139623.1
XM_006724828.3 2907 Intron XP_006724891.1

View Full Product Details