Product Details
- SNP ID
-
rs1062466
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.4:56394100 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GAAACAAACACTTTTATGTTTAAAA[T/A]TTCTTACATAAACAAAGGTTTGGGG
- Phenotype
-
MIM: 614365
MIM: 172450
- Polymorphism
- T/A, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
AASDH
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs1042040] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- AASDH
- Gene Name
- aminoadipate-semialdehyde dehydrogenase
There are no transcripts associated with this gene.
- Gene
- PPAT
- Gene Name
- phosphoribosyl pyrophosphate amidotransferase
View Full Product Details