Product Details
- SNP ID
-
rs1061814
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.4:177431525 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CATCTTCAGATAATATAAGAAAGGT[G/T]AAATAAAAATAGATAATACAATTTC
- Phenotype
-
MIM: 613228
- Polymorphism
- G/T, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
AGA
PubMed Links
Gene Details
- Gene
- AGA
- Gene Name
- aspartylglucosaminidase
View Full Product Details