Product Details

SNP ID
rs11544392
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:8288721 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTGGCAGCTGCGAAACCCAGGGAGC[T/C]GATGCCACGTGACCCAATGTGGACT
Phenotype
MIM: 607954 MIM: 610818
Polymorphism
T/C, Transition substitution
Allele Nomenclature
Literature Links
RANGRF PubMed Links

Gene Details

Gene
RANGRF
Gene Name
RAN guanine nucleotide release factor
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001177801.1 71 UTR 5 NP_001171272.1
NM_001177802.1 71 UTR 5 NP_001171273.1
NM_016492.4 71 UTR 5 NP_057576.2
XM_005256618.4 71 UTR 5 XP_005256675.1
Gene
SLC25A35
Gene Name
solute carrier family 25 member 35
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001320870.1 71 Intron NP_001307799.1
NM_001320871.1 71 Intron NP_001307800.1
NM_001320872.1 71 Intron NP_001307801.1
NM_001320877.1 71 Intron NP_001307806.1
NM_201520.2 71 UTR 3 NP_958928.1
XM_005256641.2 71 Intron XP_005256698.1
XM_011523846.1 71 Intron XP_011522148.1
XM_017024633.1 71 Intron XP_016880122.1
XM_017024634.1 71 Intron XP_016880123.1

View Full Product Details