Product Details

SNP ID
rs181244551
Assay Type
Functionally tested
NCBI dbSNP Submissions
3
Location
Chr.1:209780562 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGAGAAAGACTGGGATCTCAGAGAC[C/G]AGCTGCAAAAGAAGACTTTGCAGCT
Phenotype
MIM: 607199 MIM: 608255
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
C1orf74 PubMed Links

Gene Details

Gene
C1orf74
Gene Name
chromosome 1 open reading frame 74
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_152485.3 1842 UTR 3 NP_689698.1
Gene
IRF6
Gene Name
interferon regulatory factor 6
There are no transcripts associated with this gene.

Gene
TRAF3IP3
Gene Name
TRAF3 interacting protein 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001287754.1 1842 Intron NP_001274683.1
NM_001320143.1 1842 Missense Mutation CAG,GAG Q469E NP_001307072.1
NM_001320144.1 1842 Missense Mutation CAG,GAG Q449E NP_001307073.1
NM_025228.3 1842 Missense Mutation CAG,GAG Q469E NP_079504.2
XM_005273279.4 1842 Missense Mutation CAG,GAG Q205E XP_005273336.1
XM_005273280.2 1842 Missense Mutation CAG,GAG Q205E XP_005273337.1
XM_011510018.2 1842 Intron XP_011508320.1
XM_011510019.1 1842 Intron XP_011508321.1
XM_017002399.1 1842 Missense Mutation CAG,GAG Q449E XP_016857888.1
XM_017002400.1 1842 Intron XP_016857889.1

View Full Product Details