Product Details

SNP ID
rs181179405
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:127878309 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCAGAACGAGTATCCGTAACTGTGG[G/T]TCGTTCCTTTACTGGTGGTTGCCGG
Phenotype
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
CLRN3 PubMed Links

Gene Details

Gene
CLRN3
Gene Name
clarin 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_152311.3 503 Missense Mutation AAC,ACC N174T NP_689524.1
XM_011539274.2 503 Missense Mutation AAC,ACC N114T XP_011537576.1

View Full Product Details