Product Details

SNP ID
rs183239518
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.18:9886706 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATGGTCAGCCACACGTTCCACATGC[A/G]CACAGAGGAGTCTGATGCCTCACAG
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
TXNDC2 PubMed Links

Gene Details

Gene
TXNDC2
Gene Name
thioredoxin domain containing 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001098529.1 676 Missense Mutation CAC,CGC H76R NP_001091999.1
NM_032243.5 676 Missense Mutation CAC,CGC H9R NP_115619.4
XM_017026041.1 676 Missense Mutation CAC,CGC H9R XP_016881530.1

View Full Product Details