Product Details

SNP ID
rs183778659
Assay Type
Functionally tested
NCBI dbSNP Submissions
5
Location
Chr.1:172532537 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAACATCTAGCTCAATGGGGCAGTC[C/T]ATTGCCACAGGGAAAGGGCTTGGTG
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
SUCO PubMed Links

Gene Details

Gene
SUCO
Gene Name
SUN domain containing ossification factor
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001282750.1 189 Intron NP_001269679.1
NM_001282751.1 189 Intron NP_001269680.1
NM_014283.4 189 Intron NP_055098.1
NM_016227.3 189 Missense Mutation CCA,CTA P22L NP_057311.3
XM_006711374.2 189 Intron XP_006711437.1
XM_006711375.2 189 Intron XP_006711438.1
XM_006711376.1 189 Intron XP_006711439.1

View Full Product Details