Product Details

SNP ID
rs183720647
Assay Type
Functionally tested
NCBI dbSNP Submissions
16
Location
Chr.1:224365991 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCTGTTGCCACTTGGAATATATATC[A/G]GTGAGTATAGTTTGTTTACTTTGGT
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CNIH4 PubMed Links

Gene Details

Gene
CNIH4
Gene Name
cornichon family AMPA receptor auxiliary protein 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001277197.1 365 Missense Mutation CAT,CGT H84R NP_001264126.1
NM_001277198.1 365 Missense Mutation CAA,CGA Q84R NP_001264127.1
NM_001277199.1 365 Missense Mutation CAA,CGA Q21R NP_001264128.1
NM_001277200.1 365 Missense Mutation CAA,CGA Q84R NP_001264129.1
NM_014184.3 365 Missense Mutation CAA,CGA Q84R NP_054903.1

View Full Product Details