Product Details

SNP ID
rs184094021
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.11:17386940 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGGCTCAGGACAGGGAATCTGGAGA[A/G]ATGCTGAACTTGGGCTTGGCCTTGG
Phenotype
MIM: 600509 MIM: 600937 MIM: 613714
Polymorphism
A/G, Transition Substitution
Allele Nomenclature
Literature Links
ABCC8 PubMed Links
Additional Information
For this assay, SNP(s) [rs41282930] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.

Gene Details

Gene
ABCC8
Gene Name
ATP binding cassette subfamily C member 8
There are no transcripts associated with this gene.

Gene
KCNJ11
Gene Name
potassium voltage-gated channel subfamily J member 11
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000525.3 1349 Silent Mutation ATC,ATT I384I NP_000516.3
NM_001166290.1 1349 Silent Mutation ATC,ATT I297I NP_001159762.1
XM_006718226.3 1349 Silent Mutation ATC,ATT I297I XP_006718289.1
XM_017017680.1 1349 Silent Mutation ATC,ATT I297I XP_016873169.1
Gene
NCR3LG1
Gene Name
natural killer cell cytotoxicity receptor 3 ligand 1
There are no transcripts associated with this gene.

View Full Product Details