Product Details

SNP ID
rs184999926
Assay Type
Functionally tested
NCBI dbSNP Submissions
4
Location
Chr.1:94899473 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GATCATAAAGATGCCTCCTAGTCCC[A/G]TAAGCTGTCATACCTGCCTGGCTGG
Phenotype
MIM: 602374
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CNN3 PubMed Links

Gene Details

Gene
CNN3
Gene Name
calponin 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001286055.1 665 Silent Mutation TAC,TAT Y136Y NP_001272984.1
NM_001286056.1 665 Silent Mutation TAC,TAT Y141Y NP_001272985.1
NM_001839.4 665 Silent Mutation TAC,TAT Y182Y NP_001830.1
XM_017000245.1 665 Silent Mutation TAC,TAT Y141Y XP_016855734.1
Gene
SLC44A3
Gene Name
solute carrier family 44 member 3
There are no transcripts associated with this gene.

View Full Product Details