Product Details

SNP ID
rs185537653
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.19:11394449 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTGTTCCGGGTCCCCTCTTTCCGCC[A/G]CAGCATGAAGTCTCTGGGGGCGACT
Phenotype
MIM: 616743
Polymorphism
A/G, Transition Substitution
Allele Nomenclature
Literature Links
RGL3 PubMed Links

Gene Details

Gene
RGL3
Gene Name
ral guanine nucleotide dissociation stimulator like 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001035223.3 2168 Silent Mutation NP_001030300.3
NM_001161616.2 2168 Silent Mutation NP_001155088.2

View Full Product Details