Product Details

SNP ID
rs185706635
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:89777265 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACGGCGGCCCCAGCAACTTTGGAGC[C/T]GGCCGCAGTGTACCAGGTATGTGTG
Phenotype
MIM: 151530 MIM: 605195
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ANPEP PubMed Links

Gene Details

Gene
ANPEP
Gene Name
alanyl aminopeptidase, membrane
There are no transcripts associated with this gene.

Gene
MESP2
Gene Name
mesoderm posterior bHLH transcription factor 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001039958.1 908 Missense Mutation CCG,CTG P303L NP_001035047.1

View Full Product Details