Product Details

SNP ID
rs185879196
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:95202130 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACCTGTGCCCTTAGCCATGGAGACC[A/G]GCTGATGATAATCTTGCCCCCAACA
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ACSM6 PubMed Links

Gene Details

Gene
ACSM6
Gene Name
acyl-CoA synthetase medium-chain family member 6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_207321.2 463 Missense Mutation CAG,CGG Q113R NP_997204.2

View Full Product Details