Product Details

SNP ID
rs185854216
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:14055061 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCCCTGCTCTGGAGTGGGGATGGGG[A/C]TGGGGCATGGGCCATTGGCTTCTGA
Phenotype
MIM: 605350
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
IL27RA PubMed Links

Gene Details

Gene
IL27RA
Gene Name
interleukin 27 receptor subunit alpha
There are no transcripts associated with this gene.

Gene
PALM3
Gene Name
paralemmin 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001145028.1 566 Missense Mutation AGC,ATC S189I NP_001138500.1
XM_005259891.4 566 Missense Mutation AGC,ATC S204I XP_005259948.1
XM_011527965.2 566 Missense Mutation AGC,ATC S177I XP_011526267.1

View Full Product Details