Product Details

SNP ID
rs185922239
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:68692040 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCGTTCCGGGCTGTTCACAAACACA[C/G]AGCCCCACAGTATAACAGAAGATGT
Phenotype
MIM: 607617
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
LOC100507250 PubMed Links

Gene Details

Gene
LOC100507250
Gene Name
uncharacterized LOC100507250
There are no transcripts associated with this gene.

Gene
NUP107
Gene Name
nucleoporin 107
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
XM_005269037.3 482 Missense Mutation CAG,GAG Q126E XP_005269094.1
XM_011538576.1 482 Missense Mutation CAG,GAG Q97E XP_011536878.1

View Full Product Details