Product Details

SNP ID
rs186221155
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:154234965 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAAGATAATTCCAGTGATAGTTGTC[C/T]TTTCCATCATGATCAGAGGTCGTCT
Phenotype
MIM: 612486
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
DCHS2 PubMed Links

Gene Details

Gene
DCHS2
Gene Name
dachsous cadherin-related 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001142552.1 5616 Intron NP_001136024.1
NM_017639.3 5616 Silent Mutation AAA,AAG K2774K NP_060109.2
XM_011532045.1 5616 Silent Mutation AAA,AAG K3229K XP_011530347.1
XM_011532046.2 5616 Silent Mutation AAA,AAG K2452K XP_011530348.1
XM_011532049.2 5616 Silent Mutation AAA,AAG K1564K XP_011530351.1
XM_017008320.1 5616 Intron XP_016863809.1

View Full Product Details