Product Details

SNP ID
rs186833030
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:7329168 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGCTGGTCTTGAACTCCTGAACTCA[A/G]GTGATCCACCCGCCTCGGCCTCCCA
Phenotype
MIM: 608304
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CAGE1 PubMed Links

Gene Details

Gene
CAGE1
Gene Name
cancer antigen 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001170692.1 3003 Intron NP_001164163.1
NM_001170693.1 3003 Silent Mutation NP_001164164.1
NM_205864.2 3003 Intron NP_995586.1

View Full Product Details