Product Details

SNP ID
rs187092458
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:41180808 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCAGCACCTTGATTGCCACTTTTCC[C/T]CCAGCCACTCTGTCAAAATCAAAAC
Phenotype
MIM: 172420 MIM: 609598
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
PLCG1 PubMed Links

Gene Details

Gene
PLCG1
Gene Name
phospholipase C gamma 1
There are no transcripts associated with this gene.

Gene
ZHX3
Gene Name
zinc fingers and homeoboxes 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_015035.3 7632 UTR 3 NP_055850.1
XM_005260341.3 7632 UTR 3 XP_005260398.1
XM_005260343.4 7632 Intron XP_005260400.1
XM_006723748.3 7632 Intron XP_006723811.1
XM_006723749.3 7632 Intron XP_006723812.1
XM_011528701.2 7632 Intron XP_011527003.1
XM_011528702.2 7632 Intron XP_011527004.1
XM_011528703.2 7632 Intron XP_011527005.1
XM_011528704.2 7632 Intron XP_011527006.1
XM_011528705.2 7632 Intron XP_011527007.1
XM_011528706.2 7632 Intron XP_011527008.1
XM_011528707.2 7632 Intron XP_011527009.1
XM_011528708.2 7632 Intron XP_011527010.1
XM_011528709.2 7632 Intron XP_011527011.1
XM_011528711.2 7632 Intron XP_011527013.1
XM_011528713.2 7632 Intron XP_011527015.1
XM_011528714.2 7632 Intron XP_011527016.1
XM_011528717.2 7632 Intron XP_011527019.1
XM_011528718.2 7632 Intron XP_011527020.1
XM_011528720.1 7632 UTR 3 XP_011527022.1
XM_017027736.1 7632 Intron XP_016883225.1
XM_017027737.1 7632 UTR 3 XP_016883226.1
XM_017027738.1 7632 UTR 3 XP_016883227.1

View Full Product Details