Product Details

SNP ID
rs187102720
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:9838027 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAACAGAGGGAGGACTATTGTAAGC[A/G]GAGCCCCAGGCACTGTAGGGTCCTG
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ARPC4-TTLL3 PubMed Links

Gene Details

Gene
ARPC4-TTLL3
Gene Name
ARPC4-TTLL3 readthrough
There are no transcripts associated with this gene.

Gene
RPUSD3
Gene Name
RNA pseudouridylate synthase domain containing 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001142547.1 907 Missense Mutation CGC,TGC R334C NP_001136019.1
NM_173659.3 907 Missense Mutation CGC,TGC R349C NP_775930.2
XM_011533627.2 907 UTR 3 XP_011531929.1
Gene
TTLL3
Gene Name
tubulin tyrosine ligase like 3
There are no transcripts associated with this gene.

View Full Product Details