Product Details

SNP ID
rs188490696
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:10707549 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CATTGCAGCCCATCAGCGGCCGGAC[A/G]AGCAGAACCTCTTCGCCATTATCCA
Phenotype
MIM: 609615
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
QTRT1 PubMed Links

Gene Details

Gene
QTRT1
Gene Name
queuine tRNA-ribosyltransferase catalytic subunit 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_031209.2 605 Missense Mutation AAG,GAG K194E NP_112486.1

View Full Product Details