Product Details

SNP ID
rs188682715
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:43197034 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AACTCTGAATCTTCTTGTCCTTCTC[A/G]AAAGGACACTCTACCTGTGTCCATG
Phenotype
MIM: 614531
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
RASGEF1A PubMed Links

Gene Details

Gene
RASGEF1A
Gene Name
RasGEF domain family member 1A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001282862.1 1371 Silent Mutation TTC,TTT F438F NP_001269791.1
NM_145313.3 1371 Silent Mutation TTC,TTT F430F NP_660356.2
XM_005271809.3 1371 Silent Mutation TTC,TTT F350F XP_005271866.1
XM_011539500.2 1371 Silent Mutation TTC,TTT F350F XP_011537802.1

View Full Product Details