Product Details

SNP ID
rs189564920
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:56190202 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTCCTGTGTGTGACCTCCTGTGGAT[G/T]CTTAGCTGGGAAAAATACTTAAATG
Phenotype
MIM: 611178
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
GALP PubMed Links

Gene Details

Gene
GALP
Gene Name
galanin like peptide
There are no transcripts associated with this gene.

Gene
ZSCAN5B
Gene Name
zinc finger and SCAN domain containing 5B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001080456.2 1293 Missense Mutation AGA,AGC R371S NP_001073925.2
XM_005258864.3 1293 Missense Mutation AGA,AGC R424S XP_005258921.2
XM_006723189.3 1293 Missense Mutation AGA,AGC R371S XP_006723252.1

View Full Product Details