Product Details

SNP ID
rs189863244
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.22:41436941 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAATGGGCACGAACACCTGGGCGTC[A/C]GGGTTGAAGCTGCTCTTGATCTCCT
Phenotype
MIM: 607396
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
TOB2 PubMed Links

Gene Details

Gene
TOB2
Gene Name
transducer of ERBB2, 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_016272.3 1629 Silent Mutation NP_057356.1
XM_005261315.2 1629 Silent Mutation XP_005261372.1
XM_006724105.3 1629 Silent Mutation XP_006724168.1
XM_017028539.1 1629 Silent Mutation XP_016884028.1

View Full Product Details