Product Details

SNP ID
rs189664184
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:115831832 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCCCCACGCCTGAGACTTGCAGTAA[C/T]GTAAAACCAGTTTACCATCACTGCC
Phenotype
MIM: 601507 MIM: 609608
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
AP3S1 PubMed Links

Gene Details

Gene
AP3S1
Gene Name
adaptor related protein complex 3 sigma 1 subunit
There are no transcripts associated with this gene.

Gene
ATG12
Gene Name
autophagy related 12
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001277783.1 557 UTR 3 NP_001264712.1
NM_004707.3 557 Missense Mutation CAT,CGT H132R NP_004698.3

View Full Product Details