Product Details

SNP ID
rs190296497
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:56867108 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACCTGCATGCCCTGGCCTTTGACAG[A/C]CGGCCCAGCCACGAGATGACTGATG
Phenotype
MIM: 613395 MIM: 600968
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
MIR138-2 PubMed Links

Gene Details

Gene
MIR138-2
Gene Name
microRNA 138-2
There are no transcripts associated with this gene.

Gene
SLC12A3
Gene Name
solute carrier family 12 member 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000339.2 350 Missense Mutation AGA,AGC R107S NP_000330.2
NM_001126107.1 350 Missense Mutation AGA,AGC R106S NP_001119579.1
NM_001126108.1 350 Missense Mutation AGA,AGC R107S NP_001119580.1
XM_005256119.2 350 Missense Mutation AGA,AGC R106S XP_005256176.1

View Full Product Details