Product Details

SNP ID
rs343105
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:73554570 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TCTATGATATTAACTTTTTCTTCTA[T/C]GTAGCTAATTAATTATTCTTCTTTA
Phenotype
MIM: 605787
Polymorphism
T/C, Transition substitution
Allele Nomenclature
Literature Links
ANKRA2 PubMed Links

Gene Details

Gene
ANKRA2
Gene Name
ankyrin repeat family A member 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_023039.4 Intron NP_075526.1
XM_005248560.2 Intron XP_005248617.1
XM_017009678.1 Intron XP_016865167.1

View Full Product Details