Product Details

SNP ID
rs345632
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:13798770 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAATAGTAAAGCTAGGATTCAAGCC[C/T]CGGTTTCTCTGTTTTTTTATTTTTA
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ZSWIM4 PubMed Links

Gene Details

Gene
ZSWIM4
Gene Name
zinc finger SWIM-type containing 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_023072.2 Intron NP_075560.2
XM_005260023.3 Intron XP_005260080.1
XM_017027153.1 Intron XP_016882642.1
XM_017027154.1 Intron XP_016882643.1
XM_017027155.1 Intron XP_016882644.1
XM_017027156.1 Intron XP_016882645.1
XM_017027157.1 Intron XP_016882646.1

View Full Product Details