Product Details

SNP ID
rs5758504
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.22:41912735 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTCTCCCCAGCAGAGAAAAGGGAGT[C/T]GGGGCCATGGGAGCCTGAAGCCCAG
Phenotype
MIM: 600481
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
SHISA8 PubMed Links

Gene Details

Gene
SHISA8
Gene Name
shisa family member 8
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001207020.1 Intron NP_001193949.1
XM_006724254.3 Intron XP_006724317.1
XM_006724255.2 Intron XP_006724318.1
XM_006724256.3 Intron XP_006724319.1
Gene
SREBF2
Gene Name
sterol regulatory element binding transcription factor 2
There are no transcripts associated with this gene.

View Full Product Details