Product Details

SNP ID
rs7388583
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:102209499 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAAAACAGGTAATAAGTGTTGGTGC[A/G]GATGTGGAGAAATTAGAACCCTCAT
Phenotype
MIM: 604712
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
RRM2B PubMed Links

Gene Details

Gene
RRM2B
Gene Name
ribonucleotide reductase regulatory TP53 inducible subunit M2B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001172477.1 Intron NP_001165948.1
NM_001172478.1 Intron NP_001165949.1
NM_015713.4 Intron NP_056528.2

View Full Product Details