Product Details

SNP ID
rs7615
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:56888571 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AACAGTAAAATGAATAATATAATGA[A/G]CCCTCACACCTCCATCCCTAGTAAC
Phenotype
MIM: 600453
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
MIR3614 PubMed Links

Gene Details

Gene
MIR3614
Gene Name
microRNA 3614
There are no transcripts associated with this gene.

Gene
MTVR2
Gene Name
mouse mammary tumor virus receptor homolog 2
There are no transcripts associated with this gene.

Gene
TRIM25
Gene Name
tripartite motif containing 25
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005082.4 5082 UTR 3 NP_005073.2

View Full Product Details