Product Details
- SNP ID
-
rs7732462
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.5:172958924 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- GCCGGCCGCCGTTTAGTCCCAGAGA[C/T]AGAAAAAAGTAGAGGTAGGAGGGCG
- Phenotype
-
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
ERGIC1
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs114985587] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- ERGIC1
- Gene Name
- endoplasmic reticulum-golgi intermediate compartment 1
There are no transcripts associated with this gene.
- Gene
- LOC100268168
- Gene Name
- uncharacterized LOC100268168
There are no transcripts associated with this gene.
- Gene
- RPL26L1
- Gene Name
- ribosomal protein L26 like 1
View Full Product Details