Product Details

SNP ID
rs200994303
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:67888912 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TATACTTTTGTTCAGCAACATCTTA[C/T]GATTGGCACAGATCCTCGAACAATT
Phenotype
MIM: 604479
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
SIRT1 PubMed Links

Gene Details

Gene
SIRT1
Gene Name
sirtuin 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001142498.1 631 Intron NP_001135970.1
NM_001314049.1 631 Intron NP_001300978.1
NM_012238.4 631 Missense Mutation ACG,ATG T193M NP_036370.2

View Full Product Details