Product Details

SNP ID
rs200384144
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:70114257 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCTCTTGGTCAGCCGAACCATGAGC[C/T]GGCCCACATAGAAGCCACTGATTTG
Phenotype
MIM: 605159 MIM: 616141
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
AIFM2 PubMed Links

Gene Details

Gene
AIFM2
Gene Name
apoptosis inducing factor, mitochondria associated 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001198696.1 1257 Missense Mutation CAG,CGG Q348R NP_001185625.1
NM_032797.5 1257 Missense Mutation CAG,CGG Q348R NP_116186.1
Gene
H2AFY2
Gene Name
H2A histone family member Y2
There are no transcripts associated with this gene.

View Full Product Details