Product Details

SNP ID
rs200708795
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:43196506 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGTCTTCCAGCTGTCTTTTTCCATG[G/T]GGTTCTCGGGACCCTCACTTTCAAA
Phenotype
MIM: 614531
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
RASGEF1A PubMed Links

Gene Details

Gene
RASGEF1A
Gene Name
RasGEF domain family member 1A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001282862.1 1472 Missense Mutation CAC,CCC H472P NP_001269791.1
NM_145313.3 1472 Missense Mutation CAC,CCC H464P NP_660356.2
XM_005271809.3 1472 Missense Mutation CAC,CCC H384P XP_005271866.1
XM_011539500.2 1472 Missense Mutation CAC,CCC H384P XP_011537802.1

View Full Product Details