Product Details

SNP ID
rs200005116
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:67884758 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGCGGCCCTCGCCCTTCAGCCCGGC[G/T]GCTCCCCCTCGGCGGCGGGGGCCGA
Phenotype
MIM: 604479
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
SIRT1 PubMed Links

Gene Details

Gene
SIRT1
Gene Name
sirtuin 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001142498.1 90 Intron NP_001135970.1
NM_001314049.1 90 Intron NP_001300978.1
NM_012238.4 90 Missense Mutation GGC,TGC G13C NP_036370.2

View Full Product Details