Product Details

SNP ID
rs201774219
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:102830840 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCCCATCATCTGGCACCTCCAGGTC[A/G]AACCTCTGCAGCAGCCAGGCCATGA
Phenotype
MIM: 609300
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CYP17A1 PubMed Links

Gene Details

Gene
CYP17A1
Gene Name
cytochrome P450 family 17 subfamily A member 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000102.3 1561 Silent Mutation TTC,TTT F463F NP_000093.1
Gene
CYP17A1-AS1
Gene Name
CYP17A1 antisense RNA 1
There are no transcripts associated with this gene.

Gene
LOC107984264
Gene Name
transmembrane protein 108-like
There are no transcripts associated with this gene.

View Full Product Details