Product Details

SNP ID
rs201649411
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:70157807 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTCCACGAGGCGAGAATGATCTGCA[C/T]AGTCCACCAGAAAGACAATCCCATT
Phenotype
MIM: 607691
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
SAR1A PubMed Links

Gene Details

Gene
SAR1A
Gene Name
secretion associated Ras related GTPase 1A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001142648.1 508 Missense Mutation TAT,TGT Y102C NP_001136120.1
NM_020150.4 508 Intron NP_064535.1

View Full Product Details