Product Details

SNP ID
rs199473445
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:2445268 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCGCTCTACGCGCCCATCGCGCCCG[G/T]CGCCCCAGGTCCCGCGCCCCCTGCG
Phenotype
MIM: 607542
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
KCNQ1 PubMed Links

Gene Details

Gene
KCNQ1
Gene Name
potassium voltage-gated channel subfamily Q member 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000218.2 278 Missense Mutation GGC,GTC G57V NP_000209.2
NM_181798.1 278 Intron NP_861463.1

View Full Product Details