Product Details

SNP ID
rs200888280
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:18137955 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACCTGGATTGGAAAGTCTTATTTTG[C/T]CATGTGCATCTAGTTTCCATTTTCC
Phenotype
MIM: 607229
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
MRGPRX3 PubMed Links

Gene Details

Gene
MRGPRX3
Gene Name
MAS related GPR family member X3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_054031.3 969 Intron NP_473372.3
XM_011519882.2 969 Silent Mutation TGC,TGT C251C XP_011518184.1

View Full Product Details