Product Details

SNP ID
rs200728539
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:118344469 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGCTGTCCTCATCCTGGCTATCATT[C/T]TTCTTCAAGGTAAGGGCCTACTAGG
Phenotype
MIM: 186790 MIM: 186740
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CD3D PubMed Links

Gene Details

Gene
CD3D
Gene Name
CD3d molecule
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000732.4 126 Intron NP_000723.1
NM_001040651.1 126 Intron NP_001035741.1
XM_017018543.1 126 Intron XP_016874032.1
Gene
CD3G
Gene Name
CD3g molecule
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000073.2 126 Missense Mutation CTT,TTT L16F NP_000064.1
XM_005271724.3 126 Missense Mutation CTT,TTT L16F XP_005271781.1
XM_006718941.2 126 Missense Mutation CTT,TTT L16F XP_006719004.1

View Full Product Details