Product Details

SNP ID
rs201051174
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:55368004 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAATAAGAACAATGTGACTGAATTT[A/T]TCCTCTTAGGGCTCACACAGAACCC
Phenotype
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
OR4A15 PubMed Links

Gene Details

Gene
OR4A15
Gene Name
olfactory receptor family 4 subfamily A member 15
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001005275.1 Intron NP_001005275.1

View Full Product Details