Product Details

SNP ID
rs201006956
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:118344455 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAGGGGAAGGGCCTGGCTGTCCTCA[C/T]CCTGGCTATCATTCTTCTTCAAGGT
Phenotype
MIM: 186790 MIM: 186740
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CD3D PubMed Links

Gene Details

Gene
CD3D
Gene Name
CD3d molecule
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000732.4 112 Intron NP_000723.1
NM_001040651.1 112 Intron NP_001035741.1
XM_017018543.1 112 Intron XP_016874032.1
Gene
CD3G
Gene Name
CD3g molecule
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000073.2 112 Missense Mutation ACC,ATC T11I NP_000064.1
XM_005271724.3 112 Missense Mutation ACC,ATC T11I XP_005271781.1
XM_006718941.2 112 Missense Mutation ACC,ATC T11I XP_006719004.1

View Full Product Details