Product Details

SNP ID
rs199827989
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:17495647 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAAGGAGCTTTCGGACCGGTTGGGG[G/T]CTTTCAGCTACGGAGGAGGGAAGAG
Phenotype
MIM: 605242
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
USH1C PubMed Links

Gene Details

Gene
USH1C
Gene Name
USH1 protein network component harmonin
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001297764.1 2686 Intron NP_001284693.1
NM_005709.3 2686 Intron NP_005700.2
NM_153676.3 2686 Missense Mutation AGA,AGC R859S NP_710142.1
XM_011519832.2 2686 Intron XP_011518134.1
XM_011519834.2 2686 Intron XP_011518136.1
XM_017017072.1 2686 Intron XP_016872561.1
XM_017017073.1 2686 Intron XP_016872562.1
XM_017017074.1 2686 Intron XP_016872563.1
XM_017017075.1 2686 Missense Mutation AGA,AGC R859S XP_016872564.1

View Full Product Details