Product Details

SNP ID
rs199967551
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:66593289 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGCTTCGGATTCGGGGAACCAGGGG[A/G]CCCTCTGCACGGTGAGGGTCGAGGC
Phenotype
MIM: 603864
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CCDC87 PubMed Links

Gene Details

Gene
CCDC87
Gene Name
coiled-coil domain containing 87
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_018219.2 71 Intron NP_060689.2
Gene
CCS
Gene Name
copper chaperone for superoxide dismutase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005125.1 71 Missense Mutation ACC,GCC T10A NP_005116.1

View Full Product Details