Product Details

SNP ID
rs267606568
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:67483228 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACGTGTGATACAGGAAGGCCGAGGA[G/T]AGCTCCCGGACTTTCAAGATGGGAC
Phenotype
MIM: 605555 MIM: 608794
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
AIP PubMed Links

Gene Details

Gene
AIP
Gene Name
aryl hydrocarbon receptor interacting protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001302959.1 200 Intron NP_001289888.1
NM_001302960.1 200 Missense Mutation GAG,TAG E24* NP_001289889.1
NM_003977.3 200 Missense Mutation GAG,TAG E24* NP_003968.3
Gene
MIR6752
Gene Name
microRNA 6752
There are no transcripts associated with this gene.

Gene
PITPNM1
Gene Name
phosphatidylinositol transfer protein membrane associated 1
There are no transcripts associated with this gene.

View Full Product Details