Product Details

SNP ID
rs201436284
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:1277155 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCCATGTTGGGGAACATGTCCTGGA[G/T]GGCTTTCAGGTCCTCCTCGCTACAG
Phenotype
MIM: 606277
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
TOLLIP PubMed Links

Gene Details

Gene
TOLLIP
Gene Name
toll interacting protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001318512.1 873 Missense Mutation ATC,CTC I187L NP_001305441.1
NM_001318514.1 873 Missense Mutation ATC,CTC I168L NP_001305443.1
NM_001318515.1 873 UTR 3 NP_001305444.1
NM_001318516.1 873 Missense Mutation ATC,CTC I176L NP_001305445.1
NM_019009.3 873 Missense Mutation ATC,CTC I237L NP_061882.2
XM_017017931.1 873 Missense Mutation ATC,CTC I111L XP_016873420.1

View Full Product Details