Product Details

SNP ID
rs201791614
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:13009827 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GACGAGGCGCTGCCGCAGGGCATGC[A/T]GTGCGGGCCCCCGCAGTGCTATTGC
Phenotype
MIM: 614713
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
RASSF10 PubMed Links

Gene Details

Gene
RASSF10
Gene Name
Ras association domain family member 10
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001080521.2 405 Missense Mutation CAG,CTG Q84L NP_001073990.2

View Full Product Details