Product Details

SNP ID
rs201986043
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:13010445 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGCATCGAGCTCGACGGGTCCAGAC[C/G]GGGAGAGGAGCCAGAAGAGGTGGCG
Phenotype
MIM: 614713
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
RASSF10 PubMed Links

Gene Details

Gene
RASSF10
Gene Name
Ras association domain family member 10
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001080521.2 1023 Missense Mutation CCG,CGG P290R NP_001073990.2

View Full Product Details