Product Details
- SNP ID
-
rs200986210
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.12:48202797 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TGGTTGTCTCTGCGGAAGGGCTCTC[A/G]GAATGAAATATACTCGAGAAAGATG
- Phenotype
-
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
OR10AD1
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs17224674] are located under a probe and SNP(s) [rs75531856] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- OR10AD1
- Gene Name
- olfactory receptor family 10 subfamily AD member 1
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001004134.1 |
496 |
Nonsense Mutation |
CGA,TGA |
R166* |
NP_001004134.1 |
View Full Product Details